Tisk…

Dvojchroman amonný ≥99.5% ACS

Dodavatel: Thermo Scientific
Nebezpečí

13444.36 13444.A3
13444.36EA 2980 CZK
13444.36 13444.A3
Dvojchroman amonný ≥99.5% ACS
Ammonium dichromate

Purity: 99.5% min CAS No.: 7789-09-5 Molecular Formula: (NH4)2Cr2O7 Formula Weight: 252.10 Physical Form: Crystalline Odour: Odourless Merck Reference: 14,515 UN No.: 1439 MDL No.: MFCD00010879


Catalysts, source of pure nitrogen, in lithography, manufacturing of pigments

Vzorec: (NH₄)₂Cr₂O₇
MW: 252,06 g/mol
Bod tání: 170 °C (rozkl.)
Hustota: 2,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010879
CAS číslo: 7789-09-5
EINECS: 232-143-1
UN: 1439
ADR: 5.1,II
REACH: 01-2119661563-36-0000
Merck Index: 14,515

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR