Tisk…

Hydrogenuhličitan amonný 98

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ammonium bicarbonate

A18566.0E A18566.36
A18566.0EEA 2130 CZK
A18566.0E A18566.36
Hydrogenuhličitan amonný 98
Hydrogenuhličitan amonný

Purity: 98% CAS No.: 1066-33-7 Molecular Formula: (NH4)HCO3 Formula Weight: 79.06 Odour: Like ammonia Melting Point: ca 60° dec. Beilstein Reference: 4329606 Merck Reference: 14,497 MDL No.: MFCD00012138

Vzorec: NH₄HCO₃
MW: 79.06
Bod tání: ∼ 60 °C (rozkl.)
Hustota: 1.586
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS číslo: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR