Bromid amonný, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
380008-25G
380008-5G
SIAL380008-25GEA
6040
CZK
SIAL380008-25G
SIAL380008-5G
Bromid amonný, Sigma-Aldrich®
Bromid amonný
Vzorec:
H₄BrN MW: 97.94 Bod tání: 542 Hustota: 2.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011419 CAS číslo: 12124-97-9 EINECS: 235-183-8 Merck Index: 13,00505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...