Tisk…

Diphenylphosphine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

252964-10G 252964-50G
SIAL252964-10GEA 4290 CZK
SIAL252964-10G SIAL252964-50G
Diphenylphosphine, Sigma-Aldrich®
Diphenylphosphine
Vzorec: C₁₂H₁₁P
MW: 186.19
MDL Number: MFCD00003040
CAS číslo: 829-85-6
EINECS: 212-591-4
UN: 2845
ADR: 4.2 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR