Chlorodiphenylphosphine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
DPPC, Diphenylchlorophosphine
M-Clarity™ quality level = MQ100
C39601-100G
C39601-25G
C39601-500G
SIALC39601-100GEA
5480
CZK
SIALC39601-100G
SIALC39601-25G
SIALC39601-500G
Chlorodiphenylphosphine, Sigma-Aldrich®
Chlorodiphenylphosphine
Vzorec:
C₁₂H₁₀ClP Bod varu: 320 °C (1013 hPa) Bod tání: 14…16 °C Hustota: 1,2 g/cm³ (20 °C) Bod vzplanutí: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000529 CAS číslo: 1079-66-9 EINECS: 214-093-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...