Tisk…

Chlorodiphenylphosphine 97%, rods

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14241.09 A14241.18 A14241.30
A14241.09EA 1040 CZK
A14241.09 A14241.18 A14241.30
Chlorodiphenylphosphine 97%, rods
Chlorodiphenylphosphine

Purity: 97% CAS No.: 1079-66-9 Molecular Formula: C12H10ClP Formula Weight: 220.64 Flash Point: 138°(280°F) Refractive Index: 1.6350 Odour: Acrid Melting Point: 14-16° Boiling Point: 318-321° Beilstein Reference: 512032 UN No.: 3265 MDL No.: MFCD00000529

Vzorec: C₁₂H₁₀ClP
Bod varu: 318…321 °C
Bod tání: 14…16 °C
Hustota: 1,204
Bod vzplanutí: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS číslo: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR