Tisk…

Nitronium hexafluorophosphate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

13151.06 13151.14
13151.06EA 4770 CZK
13151.06 13151.14
Nitronium hexafluorophosphate
Nitronium hexafluorophosphate

CAS No.: 19200-21-6 Molecular Formula: NO2PF6 Formula Weight: 190.97 Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00031520

Vzorec: F₆NO₂P
Storage Temperature: Ambient
MDL Number: MFCD00031520
CAS číslo: 19200-21-6
EINECS: 242-872-7
UN: 3260
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR