Nitronium tetrafluoroborate ≥90%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Nitryl tetrafluoroborate
36374.06
36374.14
36374.06EA
1960
CZK
36374.06
36374.14
Nitronium tetrafluoroborate ≥90%
Nitronium tetrafluoroborate
Purity: 90%
CAS No.: 13826-86-3
Molecular Formula: BF4NO2
Formula Weight: 132.81
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 240° dec.
UN No.: 3260
MDL No.: MFCD00011432
Vzorec:
BF₄NO₂ MW: 132,81 g/mol Bod tání: 240 °C (rozkl.) Hustota: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00011432 CAS číslo: 13826-86-3 EINECS: 237-533-5 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...