Tisk…

Benzetoniumchlorid ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Hyamine

A16142.22EA 1620 CZK
A16142.22
Benzetoniumchlorid ≥97%
Benzetoniumchlorid
Vzorec: C₂₇H₄₂ClNO₂
MW: 448,09 g/mol
Bod tání: 157…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00011742
CAS číslo: 121-54-0
EINECS: 204-479-9
UN: 1759
ADR: 8,III
Merck Index: 14,1074

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR