Benzyltriethylammonium chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Benzyl triethylammonium chloride
M-Clarity™ quality level = MQ200
146552-100G
146552-25G
146552-500G
SIAL146552-100GEA
873
CZK
SIAL146552-100G
SIAL146552-25G
SIAL146552-500G
Benzyltriethylammonium chloride, Sigma-Aldrich®
Benzyltriethylammonium chloride
Vzorec:
C₁₃H₂₂ClN Bod tání: 190…192 °C Bod vzplanutí: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011824 CAS číslo: 56-37-1 EINECS: 200-270-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...