Tetrabutylammonium nitrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
359173-10G
359173-2G
SIAL359173-10GEA
2100
CZK
SIAL359173-10G
SIAL359173-2G
Tetrabutylammonium nitrate, Sigma-Aldrich®
Tetrabutylammonium nitrate
Vzorec:
C₁₆H₃₆N₂O₃ MW: 304,47 g/mol Bod tání: 116…120 °C |
MDL Number:
MFCD00043202 CAS číslo: 1941-27-1 EINECS: 217-726-0 UN: 1479 ADR: 5.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...