Tetramethylamoniumjodid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
235946-100G
235946-25G
SIAL235946-100GEA
2370
CZK
SIAL235946-100G
SIAL235946-25G
Tetramethylamoniumjodid, Sigma-Aldrich®
Tetramethylamoniumjodid
Vzorec:
C₄H₁₂IN Bod tání: 300 °C Hustota: 1,84 g/cm³ (20 °C) |
MDL Number:
MFCD00011629 CAS číslo: 75-58-1 EINECS: 200-881-3 UN: 2811 ADR: 6.1,III Merck Index: 12,09364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...