Tisk…

Tetraethylammonium iodide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

235938-100G 235938-25G
SIAL235938-100GEA 1890 CZK
SIAL235938-100G SIAL235938-25G
Tetraethylammonium iodide, Sigma-Aldrich®
Tetraethylammonium iodide
Vzorec: C₈H₂₀IN
Bod tání: 300 °C
Hustota: 1,566 g/cm³ (20 °C)
MDL Number: MFCD00011829
CAS číslo: 68-05-3
EINECS: 200-676-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR