Tetrabutylammonium fluoride trihydrate, Sigma-Aldrich®
Dodavatel: Merck
86872-10G
86872-250G
86872-50G
SIAL86872-10GEA
1460
CZK
SIAL86872-10G
SIAL86872-250G
SIAL86872-50G
Tetrabutylammonium fluoride trihydrate, Sigma-Aldrich®
Tetrabutylammonium fluoride trihydrate
Vzorec:
C₁₆H₃₆FN·3H₂O MW: 315,51 g/mol Bod tání: 62…63 °C |
MDL Number:
MFCD00149981 CAS číslo: 87749-50-6 EINECS: 207-057-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...