Tetraethylamoniumchlorid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetrylammonium chloride
M-Clarity™ quality level = MQ200
T2265-100G
T2265-25G
SIALT2265-100GEA
4700
CZK
SIALT2265-100G
SIALT2265-25G
Tetraethylamoniumchlorid, Sigma-Aldrich®
Tetraethylamoniumchlorid
Vzorec:
C₈H₂₀ClN MW: 165,71 g/mol Bod tání: 285 °C Hustota: 1,08 g/cm³ (20 °C) |
MDL Number:
MFCD00011828 CAS číslo: 56-34-8 EINECS: 200-267-5 Merck Index: 12,09341 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...