Tetraethylamoniumchlorid, Supelco®
Dodavatel: Merck
Synonyms:
Tetrylammonium chloride
M-Clarity™ quality level = MQ100
86616-25G
86616-5G
86616-25GEA
15340
CZK
86616-25G
86616-5G
Tetraethylamoniumchlorid, Supelco®
Tetraethylamoniumchlorid
Vzorec:
C₈H₂₀ClN MW: 165.71 Bod tání: 285 Hustota: 1.08 g/cm³ (20 °C) |
MDL Number:
MFCD00011828 CAS číslo: 56-34-8 EINECS: 200-267-5 Merck Index: 12,09341 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...