Tisk…

Tetraethylamoniumchlorid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrylammonium chloride

86614-100G 86614-25G
SIAL86614-100GEA 19080 CZK
SIAL86614-100G SIAL86614-25G
Tetraethylamoniumchlorid, Sigma-Aldrich®
Tetraethylamoniumchlorid
Vzorec: C₈H₂₀ClN
MW: 165,71 g/mol
Bod tání: 285 °C
Hustota: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00011828
CAS číslo: 56-34-8
EINECS: 200-267-5
Merck Index: 12,09341

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR