Tetrabutylammonium tribromide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide
16258.14
16258.22
16258.14EA
2710
CZK
16258.14
16258.22
Tetrabutylammonium tribromide ≥98%
Tetrabutylammonium tribromide
Purity: 98%
CAS No.: 38932-80-8
Molecular Formula: C16H36Br3N
Formula Weight: 482.18
Physical Form: Crystalline
Appearance: Yellow
Melting Point: 74-76°
Beilstein Reference: 3746114
UN No.: 3261
MDL No.: MFCD00012110
Vzorec:
C₁₆H₃₆Br₃N MW: 482.18 Bod tání: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012110 CAS číslo: 38932-80-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...