Tisk…

Tetrabutylammonium tribromide 98+%

Dodavatel: Thermo Scientific
Varování

Synonyms: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

123970250 123971000 123975000
ACRO123970250EA 2980 CZK
ACRO123970250 ACRO123971000 ACRO123975000
Tetrabutylammonium tribromide 98+%
Tetrabutylammonium tribromide
Vzorec: C₁₆H₃₆Br₃N
MW: 482,18 g/mol
Bod tání: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00012110
CAS číslo: 38932-80-8
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 71 to 75 °C
Titration Iodometric ≥98.0 %
Specific optical rotation -22° to -26° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR