Tetraoctylammonium bromide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetra-n-octylammonium bromide
M-Clarity™ quality level = MQ200
294136-25G
294136-5G
SIAL294136-25GEA
5190
CZK
SIAL294136-25G
SIAL294136-5G
Tetraoctylammonium bromide, Sigma-Aldrich®
Tetraoctylammonium bromide
Vzorec:
C₃₂H₆₈BrN Bod tání: ~ 96 °C Bod vzplanutí: 100 °C |
MDL Number:
MFCD00011863 CAS číslo: 14866-33-2 EINECS: 238-936-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...