Tisk…

Tetramethylamoniumbromid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

426296-100G 426296-25G
SIAL426296-100GEA 2320 CZK
SIAL426296-100G SIAL426296-25G
Tetramethylamoniumbromid, Sigma-Aldrich®
Tetramethylamoniumbromid
Vzorec: C₄H₁₂BrN
MW: 154.05
Bod varu: ∼360 °C (1013 hPa)
Hustota: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00011626
CAS číslo: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR