Tetrahexylammonium bromide, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
252816-25G
252816-5G
SIAL252816-25GEA
3510
CZK
SIAL252816-25G
SIAL252816-5G
Tetrahexylammonium bromide, Sigma-Aldrich®
Tetrahexylammonium bromide
Vzorec:
C₂₄H₅₂BrN Bod tání: 104 °C Hustota: 1,0036 g/cm³ (25 °C) |
MDL Number:
MFCD00011858 CAS číslo: 4328-13-6 EINECS: 224-363-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...