Tetraheptylammonium bromide ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A15218.06
A15218.14
A15218.22
A15218.06EA
1200
CZK
A15218.06
A15218.14
A15218.22
Tetraheptylammonium bromide ≥99%
Tetraheptylammonium bromide
Purity: 99%
CAS No.: 4368-51-8
Molecular Formula: [CH3(CH2)6]4NBr
Formula Weight: 490.70
Melting Point: 87-89°
MDL No.: MFCD00011861
Vzorec:
C₂₈H₆₀BrN Bod tání: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011861 CAS číslo: 4368-51-8 EINECS: 224-459-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...