Tetraethylamoniumbromid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
241059-250G
241059-50G
SIAL241059-250GEA
5570
CZK
SIAL241059-250G
SIAL241059-50G
Tetraethylamoniumbromid, Sigma-Aldrich®
Tetraethylamoniumbromid
Vzorec:
C₈H₂₀BrN Bod tání: > 285 °C Hustota: 1,43 g/cm³ (20 °C) |
MDL Number:
MFCD00011825 CAS číslo: 71-91-0 EINECS: 200-769-4 Merck Index: 13,09274 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...