Tisk…

Tetraethylamoniumbromid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrylammonium bromide

M-Clarity™ quality level = MQ200

140023-1KG 140023-250G
SIAL140023-1KGEA 4890 CZK
SIAL140023-1KG SIAL140023-250G
Tetraethylamoniumbromid, Sigma-Aldrich®
Tetraethylamoniumbromid
Vzorec: C₈H₂₀BrN
Bod tání: > 285 °C
Hustota: 1,43 g/cm³ (20 °C)
MDL Number: MFCD00011825
CAS číslo: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR