Tisk…

Tetradodecylammonium bromide, Sigma-Aldrich®

Dodavatel: Merck
Varování

87249-25G 87249-5G
SIAL87249-25GEA 16630 CZK
SIAL87249-25G SIAL87249-5G
Tetradodecylammonium bromide, Sigma-Aldrich®
Tetradodecylammonium bromide
Vzorec: C₄₈H₁₀₀BrN
CAS číslo: 14866-34-3
EINECS: 238-937-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR