Tisk…

Tetrabutylphosphonium bromide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

189138-100G 189138-500G
SIAL189138-100GEA 1850 CZK
SIAL189138-100G SIAL189138-500G
Tetrabutylphosphonium bromide, Sigma-Aldrich®
Tetrabutylphosphonium bromide
Vzorec: C₁₆H₃₆BrP
Bod tání: 95…101 °C
MDL Number: MFCD00011853
CAS číslo: 3115-68-2
EINECS: 221-487-8
UN: 3464
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR