Cetrimonium bromid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
M-Clarity™ quality level = MQ200
52370-100G
52370-500G
SIAL52370-100GEA
1770
CZK
SIAL52370-100G
SIAL52370-500G
Cetrimonium bromid, Sigma-Aldrich®
Cetrimonium bromid
Vzorec:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ MW: 364.45 Bod tání: 237…243 °C Bod vzplanutí: 244 °C (ot. pohárek) Storage Temperature: Ambient |
MDL Number:
MFCD00011772 CAS číslo: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9 III Merck Index: 13,02034 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...