Tetraethylammonium tetrafluoroborate for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tetrylammonium tetrafluoroborate
M-Clarity™ quality level = MQ200
8.18246.0025
8.18246.0100
8.18246.0025EA
2490
CZK
8.18246.0025
8.18246.0100
Tetraethylammonium tetrafluoroborate for synthesis, Sigma-Aldrich®
Tetraethylammonium tetrafluoroborate
Vzorec:
C₈H₂₀BF₄N MW: 217,06 g/mol Bod tání: 365…368 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011827 CAS číslo: 429-06-1 EINECS: 207-055-1 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Solubility | (20 °C) soluble |
Melting Point | 365 - 368 °C |
Molar Mass | 217.06 g/mol |
Bulk Density | 400 kg/m3 |
Hill Formula | C8H20BF4N |
Chemical Formula | (C2H5)4NBF4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...