Tisk…

Tetrabutylammonium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

217964-100G 217964-25G 217964-5G
SIAL217964-100GEA 8030 CZK
SIAL217964-100G SIAL217964-25G SIAL217964-5G
Tetrabutylammonium tetrafluoroborate, Sigma-Aldrich®
Tetrabutylammonium tetrafluoroborate
Vzorec: C₁₆H₃₆BF₄N
Bod tání: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS číslo: 429-42-5
EINECS: 207-058-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR