Trioctylphosphine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
TOP, Tri-n-octylphosphine
117854-100ML
117854-4X25ML
117854-500ML
SIAL117854-100MLEA
6520
CZK
SIAL117854-100ML
SIAL117854-4X25ML
SIAL117854-500ML
Trioctylphosphine, Sigma-Aldrich®
Trioctylphosphine
Vzorec:
C₂₄H₅₁P Bod varu: 150…155 °C (0,03 mmHg) Bod tání: 48 °C Hustota: 0,831 g/cm³ (20 °C) |
MDL Number:
MFCD00015298 CAS číslo: 4731-53-7 EINECS: 225-234-2 UN: 1760 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...