(Ethyl)triphenylphosphonium bromide, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Ethyltriphenylphosphonium bromide
M-Clarity™ quality level = MQ200
E50604-100G
E50604-25G
SIALE50604-100GEA
3270
CZK
SIALE50604-100G
SIALE50604-25G
(Ethyl)triphenylphosphonium bromide, Sigma-Aldrich®
(Ethyl)triphenylphosphonium bromide
Vzorec:
C₂₀H₂₀BrP MW: 371,26 g/mol Bod tání: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011838 CAS číslo: 1530-32-1 EINECS: 216-223-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...