(1-Octyl)triphenylphosphonium bromide ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Octyltriphenylphosphonium Bromide
L02412.06
L02412.14
L02412.22
L02412.06EA
999
CZK
L02412.06
L02412.14
L02412.22
(1-Octyl)triphenylphosphonium bromide ≥97%
(1-Octyl)triphenylphosphonium bromide
Purity: 97%
CAS No.: 42036-78-2
Molecular Formula: C26H32BrP
Formula Weight: 455.42
Melting Point: 61-63°
Beilstein Reference: 3642460
Vzorec:
H₃C(CH₂)₇P(C₆H₅)₃Br MW: 455,42 g/mol Bod tání: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051874 CAS číslo: 42036-78-2 EINECS: 255-635-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...