Tisk…

Dimethyl methylphosphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

64258-1L-F 64258-250ML-F
SIAL64258-1L-FEA 7090 CZK
SIAL64258-1L-F SIAL64258-250ML-F
Dimethyl methylphosphonate, Sigma-Aldrich®
Dimethyl methylphosphonate
Vzorec: C₃H₉O₃P
Bod varu: 181 °C (1013 hPa)
Hustota: 1,1596 g/cm³ (20 °C)
MDL Number: MFCD00008349
CAS číslo: 756-79-6
EINECS: 212-052-3
Merck Index: 13,03425

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR