Tisk…

Arsenic (II) sulphide tech. 90%, technická kvalita

Dodavatel: Thermo Fisher Scientific
Nebezpečí

17284.14 17284.06
17284.14EA 6700 CZK
17284.14 17284.06
Arsenic (II) sulphide tech. 90%, technická kvalita
Arsenic (II) sulphide

Purity: 90% CAS No.: 12044-79-0 Molecular Formula: As2S2 Formula Weight: 213.97 Physical Form: Powder Density: 3.5 Melting Point: ≈307° Boiling Point: 565° UN No.: 1557 MDL No.: MFCD00049841

Vzorec: As₂S₂
Bod varu: 565 °C
Bod tání: ∼307 °C
Hustota: 3,5
Storage Temperature: Ambient
MDL Number: MFCD00049841
CAS číslo: 1303-32-8
UN: 1557
ADR: 6.1,III
Merck Index: 12,00834

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR