Tisk…

Triphenyl phosphite ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A18662.0E A18662.36
A18662.0EEA 4090 CZK
A18662.0E A18662.36
Triphenyl phosphite ≥97%
Triphenyl phosphite

Purity: 97% CAS No.: 101-02-0 Molecular Formula: C18H15O3P Formula Weight: 310.29 Flash Point: 146°(295°F) Odour: Mild Boiling Point: 360° UN No.: 3077 MDL No.: MFCD00003032

Vzorec: C₁₈H₁₅O₃P
Bod varu: 360 °C
Bod tání: 22…24 °C
Hustota: 1,184
Bod vzplanutí: 146 °C (295 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS číslo: 101-02-0
EINECS: 202-908-4
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR