Tisk…

Trimethyl phosphite ≥97%

Dodavatel: Thermo Scientific
Varování

30132.22 30132.36 30132.A4
30132.22EA 1870 CZK
30132.22 30132.36 30132.A4
Trimethyl phosphite ≥97%
Trimethyl phosphite

Purity: 97% CAS No.: 121-45-9 Molecular Formula: C3H9O3P Formula Weight: 124.08 Flash Point: 27°(80°F) Physical Form: Liquid Density: 1.052 Refractive Index: 1.4080 Sensitivity: Air and Moisture Sensitive Appearance: Colourless Odour: Unpleasant Melting Point: -78° Boiling Point: 111-112° UN No.: 2329 MDL No.: MFCD00008350

Vzorec: C₃H₉O₃P
Bod varu: 111…112 °C
Bod tání: –78 °C
Hustota: 1,052
Bod vzplanutí: 27 °C (80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008350
CAS číslo: 121-45-9
EINECS: 204-471-5
UN: 2329
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR