Triisopropyl phosphite, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tri-i-propylphosphite
M-Clarity™ quality level = MQ200
T67806-100ML
T67806-500ML
SIALT67806-100MLEA
1590
CZK
SIALT67806-100ML
SIALT67806-500ML
Triisopropyl phosphite, Sigma-Aldrich®
Triisopropyl phosphite
Vzorec:
C₉H₂₁O₃P Bod varu: 63 °C (1013 hPa) Hustota: 0,9361 g/cm³ (0 °C) |
MDL Number:
MFCD00008874 CAS číslo: 116-17-6 EINECS: 204-130-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...