Tisk…

Triisopropyl phosphite ≥90%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tri-i-propylphosphite

A15247.AE A15247.AK A15247.AP
A15247.AEEA 1500 CZK
A15247.AE A15247.AK A15247.AP
Triisopropyl phosphite ≥90%
Triisopropyl phosphite

Purity: 90+% CAS No.: 116-17-6 Molecular Formula: [(CH3)2CHO]3P Formula Weight: 208.24 Flash Point: 46°(114°F) Refractive Index: 1.4110 Odour: Characteristic Boiling Point: 63-64°/11mm Beilstein Reference: 1701528 UN No.: 1993 MDL No.: MFCD00008874

Vzorec: C₉H₂₁O₃P
MW: 208.24
Bod varu: 63…64 °C (11 mmHg)
Hustota: 0.908
Bod vzplanutí: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008874
CAS číslo: 116-17-6
EINECS: 204-130-0
UN: 1992
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR