Trimethyl phosphate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
132195-1KG
132195-250G
SIAL132195-1KGEA
3580
CZK
SIAL132195-1KG
SIAL132195-250G
Trimethyl phosphate, Sigma-Aldrich®
Trimethyl phosphate
Vzorec:
C₃H₉O₄P Bod varu: 197,2 °C (1013 hPa) Bod tání: –46 °C Hustota: 1,21 g/cm³ (20 °C) Bod vzplanutí: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008348 CAS číslo: 512-56-1 EINECS: 208-144-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...