Diethyl phosphonate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Packaged under argon.
44000.14
44000.22
44000.36
44000.A4
44000.14EA
1930
CZK
44000.14
44000.22
44000.36
44000.A4
Diethyl phosphonate ≥97%
Diethyl phosphonate
Purity: 97+%
CAS No.: 762-04-9
Molecular Formula: (C2H5O)2P(O)H
Formula Weight: 138.10
Flash Point: 74°(165°F)
Physical Form: Liquid, Packaged under argon
Density: 1.070
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Weak
Melting Point: -70°
Boiling Point: 50-51°/2mm
MDL No.: MFCD00044573
Vzorec:
C₄H₁₁O₃P Bod varu: 50…51 °C (2 mmHg) Bod tání: –70 °C Hustota: 1,070 Bod vzplanutí: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044573 CAS číslo: 762-04-9 EINECS: 212-091-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...