Arsenic (V) oxide ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Arsenic pentoxide
14668.14
14668.22
14668.14EA
4640
CZK
14668.14
14668.22
Arsenic (V) oxide ≥99.9% (metals basis)
Arsenic (V) oxide
Purity: 99.9%
CAS No.: 1303-28-2
Molecular Formula: As2O5
Formula Weight: 229.79
Physical Form: -100 Mesh Powder
Density: 4.32
Appearance: White
Odour: Odourless
Melting Point: 315° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,798
UN No.: 1559
MDL No.: MFCD00003434
Vzorec:
As₂O₅ MW: 229.84 Bod tání: 315 °C (rozkl.) Hustota: 4.32 Storage Temperature: Ambient |
MDL Number:
MFCD00003434 CAS číslo: 1303-28-2 EINECS: 215-116-9 UN: 1559 ADR: 6.1 II Merck Index: 14,798 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...