Tisk…

Oxid antimonitý ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

A11123.0E A11123.22 A11123.36
A11123.0EEA 7590 CZK
A11123.0E A11123.22 A11123.36
Oxid antimonitý ≥99%
Oxid antimonitý

Purity: 99% CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Odour: Odourless Melting Point: 656° Boiling Point: 1550° subl. Merck Reference: 14,711 MDL No.: MFCD00011214

Vzorec: O₃Sb₂
MW: 291.52
Bod varu: 1550 °C (subl.)
Bod tání: 656 °C
Hustota: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS číslo: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1 III
Merck Index: 13,00717

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR