Tisk…

Oxid antimonitý ≥99.9% (metals basis)

Dodavatel: Thermo Scientific
Varování

36314.18 36314.30
36314.18EA 6220 CZK
36314.18 36314.30
Oxid antimonitý ≥99.9% (metals basis)
Antimony (III) oxide

Purity: 99.9% CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Physical Form: 25mm and down Sintered Lump Density: 5.67 Appearance: White Odour: Odourless Melting Point: 656° Boiling Point: 1550° subl. Merck Reference: 14,711 MDL No.: MFCD00011214

Vzorec: O₃Sb₂
Bod varu: 1550 °C (subl.)
Bod tání: 656 °C
Hustota: 5,67
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS číslo: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1,III
Merck Index: 14,711

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR