Tisk…

Bismuth(III) nitrate, hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Bismuth trinitrate

10657.14 10657.22
10657.14EA 3410 CZK
10657.14 10657.22
Bismuth(III) nitrate, hydrate ≥99.999% (metals basis), Puratronic®
Bismuth(III) nitrate

It is used in electronic ceramics and fluorescent paints. It is employed as an oxidant for selective oxidation of sulfides to sulfoxides, as a nitrating reagent for synthesizing nitro aromatic compounds, and as a precursor for Dragendorff reagent for use in TLC stain and for testing the presence of alkaloids. On silica gel support, it has been reported to be an efficient catalyst for the Biginelli reaction.


Purity: 99.999% CAS No.: 10361-44-1 Molecular Formula: Bi(NO3)3.xH2O Formula Weight: 394.99(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Colourless to white Melting Point: 30° dec. Merck Reference: 14,1271 UN No.: 1477 MDL No.: MFCD00003460

Vzorec: Bi(NO₃)₃·xH₂O
MW: 485,07 g/mol
Bod varu: 75…80 °C (1013 hPa)
Bod tání: 30 °C (rozkl.)
Hustota: 2,736 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003460
CAS číslo: 10361-44-1
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Merck Index: 14,1271

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR