Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Bismuth triiodide
Ampuled under argon
35681.06
35681.14
35681.06EA
8310
CZK
35681.06
35681.14
Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Bismuth(III) iodide
Purity: 99.998%
CAS No.: 7787-64-6
Molecular Formula: BiI3
Formula Weight: 589.69
Physical Form: -10 Mesh Granules, Ampouled under argon
Density: 5.778
Melting Point: 408°
Boiling Point: ca 500°
Merck Reference: 14,1269
UN No.: 3260
MDL No.: MFCD00010894
Vzorec:
BiI₃ Bod varu: ∼500 °C Bod tání: 408 °C Hustota: 5,778 Storage Temperature: Ambient |
MDL Number:
MFCD00010894 CAS číslo: 7787-64-6 EINECS: 232-127-4 UN: 3260 ADR: 8,III Merck Index: 14,1269 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...