Arsenic(III) iodide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
45067.18
45067.09
45067.18EA
17780
CZK
45067.18
45067.09
Arsenic(III) iodide ≥98%
Arsenic(III) iodide
Purity: 98%
CAS No.: 7784-45-4
Molecular Formula: AsI3
Formula Weight: 455.64
Physical Form: Small Pieces
Density: 4.39
Melting Point: 140°
Boiling Point: 424°
Merck Reference: 14,803
UN No.: 1557
MDL No.: MFCD00014170
Vzorec:
AsI₃ Bod varu: 424 °C Bod tání: 140 °C Hustota: 4,39 Storage Temperature: Ambient |
MDL Number:
MFCD00014170 CAS číslo: 7784-45-4 EINECS: 232-068-4 UN: 1557 ADR: 6.1,II Merck Index: 14,803 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...