Tisk…

Chlorid antimonitý, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Antimony trichloride

Ampuled under argon

43449.22 43449.14
43449.22EA 24920 CZK
43449.22 43449.14
Chlorid antimonitý, ultra dry ≥99.999% (metals basis)
Chlorid antimonitý

Purity: 99.999% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Physical Form: Crystals, Ampouled under argon Density: 3.14 Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Vzorec: SbCl₃
MW: 228.12
Bod varu: 223.5 °C
Bod tání: 73.4 °C
Hustota: 3.14
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS číslo: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 14,707

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR