Phosphoryl tribromide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Phosphorus(V) oxybromide, Phosphorus (V) oxide bromide, Phosphoryl bromide, Phosphorus oxybromide
Purity: 98% min
CAS No.: 7789-59-5
Molecular Formula: POBr3
Formula Weight: 286.73
Physical Form: Lump
Density: 2.822
Melting Point: 56°
Boiling Point: 189°
Solubility: Decomposes in water. Soluble in ether, benzene, and sulfuric acid
Merck Reference: 14,7348
UN No.: 1939
MDL No.: MFCD00036298
As a chemical intermediate
Vzorec:
Br₃OP MW: 286.69 Bod varu: 189 °C Bod tání: 56 °C Hustota: 2.822 Storage Temperature: Ambient |
MDL Number:
MFCD00036298 CAS číslo: 7789-59-5 EINECS: 232-177-7 UN: 1939 ADR: 8 II Merck Index: 12,07505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...