Bismuth(III) bromide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
654981-1G
654981-5G
SIAL654981-1GEA
3610
CZK
SIAL654981-1G
SIAL654981-5G
Bismuth(III) bromide, Sigma-Aldrich®
Bismuth(III) bromide
Vzorec:
BiBr₃ MW: 448,69 g/mol Bod varu: 278 °C (11 mmHg) Bod tání: 218 °C Hustota: 5,72 g/cm³ (20 °C) |
MDL Number:
MFCD00014195 CAS číslo: 7787-58-8 EINECS: 232-121-1 UN: 3260 ADR: 8,III Merck Index: 12,01299 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...